Product Details

SNP ID
rs13479653
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.8:24852623 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCCACAAAAATCCCACAAATTTCT[A/G]GCAGTGCAATGATTAGAACATAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Adam32 PubMed Links

Gene Details

Gene
Adam32
Gene Name
a disintegrin and metallopeptidase domain 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153397.2 Intron NP_700446.2

View Full Product Details