Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLCE
Entrez Gene ID
102130661
Gene Name
glucuronic acid epimerase
Gene Aliases
-
Chromosome Location
Chr.7: 46907614 - 47031413 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005559917.2 XP_005559974.1 2 - 3 278 134

View Full Product Details