Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRN1
Entrez Gene ID
102120690
Gene Name
leucine rich repeat neuronal 1
Gene Aliases
-
Chromosome Location
Chr.2: 134693440 - 134739093 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005547659.2 XP_005547716.1 1 - 2 511 77

View Full Product Details