Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTN1
Entrez Gene ID
102144558
Gene Name
histatin 1
Gene Aliases
Histatin-1
Chromosome Location
Chr.5: 64538301 - 64543450 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005595983.2 XP_005596040.1 3 - 4 142 87

View Full Product Details