Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cybb
Entrez Gene ID
13058
Gene Name
cytochrome b-245, beta polypeptide
Gene Aliases
C88302, CGD91-phox, Cgd, Cyd, Nox2, gp91-1, gp91phox
Chromosome Location
Chr.X: 9435254 - 9487766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007807.5 NP_031833.3 2 - 3 218 149
GenBank mRNA AK033570.1 - 2 - 3 162 149
AK053920.1 - 2 - 3 162 149
AK079926.1 - 2 - 3 165 149
AK152257.1 - 2 - 3 189 149
AK153154.1 - 2 - 3 189 149
AK153259.1 - 2 - 3 189 149
AK154126.1 - 2 - 3 218 149
AK171526.1 - 2 - 3 204 149
BC042838.1 - 2 - 3 185 149
BC071229.1 - 2 - 3 173 149
BY194282.1 - 2 - 3 218 149
FJ168469.1 - 2 - 3 207 149
U43384.1 - 2 - 3 144 149

View Full Product Details