Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cybb
Entrez Gene ID
13058
Gene Name
cytochrome b-245, beta polypeptide
Gene Aliases
C88302, CGD91-phox, Cgd, Cyd, Nox2, gp91-1, gp91phox
Chromosome Location
Chr.X: 9435254 - 9487766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007807.5 NP_031833.3 9 - 10 1215 69
GenBank mRNA AK033570.1 - 9 - 10 1160 69
AK053920.1 - 9 - 10 1159 69
AK152257.1 - 9 - 10 1186 69
AK153071.1 - 2 - 3 325 69
AK153154.1 - 9 - 10 1186 69
AK153259.1 - 9 - 10 1186 69
AK160031.1 - 4 - 5 617 69
AK171526.1 - 9 - 10 1201 69
BC042838.1 - 9 - 10 1182 69
BC071229.1 - 9 - 10 1170 69
FJ168469.1 - 9 - 10 1204 69
U43384.1 - 9 - 10 1141 69

View Full Product Details