The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038700.2 | NP_001033789.1 | 1 - 2 | 230 | 120 |
NM_001177648.1 | NP_001171119.1 | 1 - 2 | 230 | 120 | |
NM_001177649.1 | NP_001171120.1 | 1 - 2 | 230 | 120 | |
NM_019406.3 | NP_062279.1 | 1 - 2 | 230 | 120 | |
GenBank mRNA | AK122308.1 | - | 1 - 2 | 230 | 120 |
AK157135.1 | - | 1 - 2 | 221 | 120 | |
AK170497.1 | - | 1 - 2 | 180 | 120 | |
AK172100.1 | - | 1 - 2 | 174 | 120 | |
BC003867.1 | - | 1 - 2 | 161 | 120 |