The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033541.4 | NP_291019.1 | 6 - 7 | 1420 | 88 |
GenBank mRNA | AB067528.1 | - | 5 - 6 | 1077 | 88 |
AF459815.1 | - | 5 - 6 | 1077 | 88 | |
AF459816.1 | - | 5 - 6 | 1077 | 88 | |
AK028986.1 | - | 6 - 7 | 1421 | 88 | |
AK078952.1 | - | 5 - 6 | 1088 | 88 | |
AK163253.1 | - | 5 - 6 | 1093 | 88 | |
BC108955.1 | - | 5 - 6 | 1086 | 88 | |
BC126877.1 | - | 5 - 6 | 1085 | 88 |