Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcm6
Entrez Gene ID
17219
Gene Name
minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S. cerevisiae)
Gene Aliases
ASP-l1, D1Wsu22e, Mcmd6
Chromosome Location
Chr.1: 128331591 - 128359656 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008567.1 NP_032593.1 11 - 12 1721 79
GenBank mRNA AK078126.1 - 5 - 6 715 79
AK088493.1 - 11 - 12 1720 79
AK145520.1 - 11 - 12 1712 79
AK146272.1 - 11 - 12 1718 79
AK146770.1 - 11 - 12 1714 79
AK146896.1 - 11 - 12 1596 79
AK150726.1 - 11 - 12 1669 79
BC050886.1 - 11 - 12 1729 79
BC051658.1 - 11 - 12 1696 79
BC057584.1 - 11 - 12 1699 79
D86726.1 - 11 - 12 1721 79

View Full Product Details