The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008516.4 | NP_032542.1 | 1 - 2 | 512 | 68 |
GenBank mRNA | AK139355.1 | - | 1 - 2 | 470 | 68 |
AK139965.1 | - | 1 - 2 | 513 | 68 | |
AK160841.1 | - | 1 - 2 | 391 | 68 | |
AK164849.1 | - | 1 - 2 | 471 | 68 | |
BC058701.1 | - | 1 - 2 | 445 | 68 | |
D45913.1 | - | 1 - 2 | 458 | 68 |