The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010499.4 | NP_034629.3 | 1 - 1 | 1451 | 82 |
GenBank mRNA | AK032692.1 | - | - | 1449 | 82 |
AK144601.1 | - | 1 - 1 | 1448 | 82 | |
AK150860.1 | - | 1 - 1 | 1446 | 82 | |
AK152747.1 | - | 1 - 1 | 1446 | 82 | |
AK170433.1 | - | - | 1448 | 82 | |
AK172332.1 | - | 1 - 1 | 1391 | 82 | |
BC002067.1 | - | 1 - 1 | 1436 | 82 | |
BP770574.1 | - | 1 - 1 | 82 | ||
M31042.1 | - | 1 - 1 | 1448 | 82 | |
M59821.1 | - | 1 - 1 | 1451 | 82 |