The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025813.3 | NP_080089.1 | 15 - 16 | 1440 | 64 |
GenBank mRNA | AK004586.1 | - | 15 - 16 | 1441 | 64 |
AK032688.1 | - | 15 - 16 | 1408 | 64 | |
AK075859.1 | - | 15 - 16 | 1470 | 64 | |
AK135235.1 | - | 15 - 16 | 1441 | 64 | |
AK136108.1 | - | 15 - 16 | 1432 | 64 | |
AK152444.1 | - | 15 - 16 | 1462 | 64 | |
AK159554.1 | - | 15 - 16 | 1468 | 64 | |
AK166645.1 | - | 9 - 10 | 832 | 64 | |
AK191801.1 | - | 5 - 6 | 372 | 64 | |
AK214765.1 | - | 4 - 5 | 238 | 64 | |
BC024891.1 | - | 15 - 16 | 1422 | 64 |