Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gmppa
Entrez Gene ID
69080
Gene Name
GDP-mannose pyrophosphorylase A
Gene Aliases
1810012N01Rik
Chromosome Location
Chr.1: 75435912 - 75443176 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133708.3 NP_598469.1 5 - 6 515 62
GenBank mRNA AK007463.1 - 5 - 6 506 62
AK144311.1 - 5 - 6 780 62
AK150473.1 - 6 - 7 651 62
AK184824.1 - 1 - 2 78 62
AK189243.1 - 1 - 2 78 62
AK190069.1 - 1 - 2 78 62
BC008116.1 - 5 - 6 474 62

View Full Product Details