Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Afap1l2
Entrez Gene ID
226250
Gene Name
actin filament associated protein 1-like 2
Gene Aliases
AU041783, C86904, mKIAA1914
Chromosome Location
Chr.19: 56912354 - 57008590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177796.1 NP_001171267.1 18 - 19 2785 96
NM_001177797.1 NP_001171268.1 17 - 18 2731 96
NM_146102.2 NP_666214.1 19 - 20 2837 96
GenBank mRNA AK048649.1 - 19 - 20 2491 96
AK087449.1 - 18 - 19 2422 96
AK220430.1 - 17 - 18 2354 96
BC031515.1 - 18 - 19 2387 96

View Full Product Details