Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp11
Entrez Gene ID
22648
Gene Name
zinc finger protein 11
Gene Aliases
6720465D24, Krox-6, Zfp-11
Chromosome Location
Chr.5: 129654595 - 129670088 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172462.4 NP_766050.1 2 - 3 127 92
GenBank mRNA AK032872.1 - 2 - 3 141 92
AK085077.1 - 1 - 2 113 92
BB623389.1 - 2 - 3 141 92
BC058553.1 - 2 - 3 125 92
BC094590.1 - 1 - 2 112 92
BC139312.1 - 1 - 2 107 92
BC139315.1 - 1 - 2 107 92

View Full Product Details