Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amz1
Entrez Gene ID
231842
Gene Name
archaelysin family metallopeptidase 1
Gene Aliases
5330426I05, 6530401C20Rik
Chromosome Location
Chr.5: 140724127 - 140753312 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173405.2 NP_775581.1 7 - 8 1303 66
GenBank mRNA AJ879912.1 - 5 - 6 944 66
AK078332.1 - 7 - 8 1303 66
AK155136.1 - - 1190 66
AK170176.1 - 6 - 7 1277 66
BC046428.1 - 2 - 3 271 66
BC129841.1 - 5 - 6 1086 66
BC129842.1 - 5 - 6 1038 66

View Full Product Details