The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025600.1 | NP_001020771.1 | 1 - 2 | 154 | 69 |
NM_018770.3 | NP_061240.3 | 1 - 2 | 154 | 69 | |
NM_207675.2 | NP_997558.2 | 1 - 2 | 154 | 69 | |
NM_207676.2 | NP_997559.1 | 1 - 2 | 154 | 69 | |
GenBank mRNA | AB052293.1 | - | 1 - 2 | 154 | 69 |
AB064265.2 | - | 1 - 2 | 133 | 69 | |
AB092414.1 | - | 1 - 2 | 133 | 69 | |
AB183399.1 | - | 1 - 2 | 133 | 69 | |
AB183400.1 | - | 1 - 2 | 133 | 69 | |
AB183401.1 | - | 1 - 2 | 133 | 69 | |
AB183402.1 | - | 1 - 2 | 133 | 69 | |
AB377192.1 | - | 1 - 2 | 154 | 69 | |
AF434663.1 | - | 1 - 2 | 249 | 69 | |
AF539424.1 | - | 1 - 2 | 146 | 69 | |
AK013775.1 | - | 1 - 2 | 155 | 69 | |
AK013911.1 | - | 1 - 2 | 248 | 69 | |
AK053839.1 | - | 1 - 2 | 155 | 69 | |
AK172628.1 | - | 1 - 2 | 157 | 69 | |
AY351388.1 | - | 1 - 2 | 133 | 69 | |
BC057125.1 | - | 1 - 2 | 142 | 69 | |
BC090970.1 | - | 1 - 2 | 333 | 69 | |
BC095986.1 | - | 1 - 2 | 146 | 69 | |
DQ279856.1 | - | 1 - 2 | 133 | 69 | |
JQ782219.1 | - | 1 - 2 | 133 | 69 |