Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nobox
Entrez Gene ID
18291
Gene Name
NOBOX oogenesis homeobox
Gene Aliases
OG2, Og2x
Chromosome Location
Chr.6: 43303674 - 43317378 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130869.3 NP_570939.1 4 - 5 567 73
GenBank mRNA AB221582.1 - 4 - 5 544 73
AY061761.1 - 4 - 5 575 73
BC125280.1 - 4 - 5 563 73
BC125282.1 - 4 - 5 563 73

View Full Product Details