Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajb11
Entrez Gene ID
67838
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 11
Gene Aliases
1810031F23Rik, ABBP-2, AL024055, Dj9, ERdj3, ERj3p
Chromosome Location
Chr.16: 22857845 - 22879634 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190804.1 NP_001177733.1 1 - 2 270 80
NM_001190805.1 NP_001177734.1 1 - 2 270 80
NM_026400.5 NP_080676.3 1 - 2 270 80
GenBank mRNA AK010861.1 - 1 - 2 242 80
AK050500.1 - 1 - 2 229 80
AK075785.1 - 1 - 2 252 80
AK075953.1 - 1 - 2 232 80
AK148511.1 - 1 - 2 250 80
AK166730.1 - 1 - 2 228 80
AY054981.1 - 1 - 2 146 80
BC003999.2 - 1 - 2 201 80
BC018282.1 - 1 - 2 197 80
BC040747.1 - 1 - 2 201 80
BY270762.1 - 1 - 2 271 80

View Full Product Details