Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc6a8
Entrez Gene ID
102857
Gene Name
solute carrier family 6 (neurotransmitter transporter, creatine), member 8
Gene Aliases
AA589632, CRT, CRTR, CT1, CTR5, Creat
Chromosome Location
Chr.X: 73673133 - 73682502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142809.1 NP_001136281.1 6 - 7 1670 63
NM_001142810.1 NP_001136282.1 6 - 7 1670 63
NM_133987.2 NP_598748.1 6 - 7 1670 63
GenBank mRNA AB077327.1 - 6 - 7 1018 63
AF459435.1 - 6 - 7 1653 63
AJ748650.1 - 3 - 4 260 63
BC024444.1 - 5 - 6 683 63
BC029000.1 - 4 - 5 433 63
BC049801.1 - 6 - 7 845 63
BC141067.1 - 7 - 8 1411 63
BC145319.1 - 7 - 8 1075 63
EU523148.1 - 2 - 3 719 63
KC994641.1 - 2 - 3 719 63

View Full Product Details