The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081074.1 | NP_001074543.1 | 9 - 10 | 1488 | 118 |
GenBank mRNA | AY566862.1 | - | 9 - 10 | 1485 | 118 |
AY566863.1 | - | 9 - 10 | 1461 | 118 | |
BC038942.1 | - | 7 - 8 | 1164 | 118 |