Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oasl2
Entrez Gene ID
23962
Gene Name
2'-5' oligoadenylate synthetase-like 2
Gene Aliases
M1204, Mmu-OASL, Oasl
Chromosome Location
Chr.5: 114896934 - 114912245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011854.2 NP_035984.2 4 - 5 1635 72
GenBank mRNA AF068835.1 - 4 - 5 1621 72
AK010034.1 - 4 - 5 977 72
AK150080.1 - 2 - 3 426 72
AK150492.1 - 4 - 5 966 72
AK157649.1 - 4 - 5 1003 72
AK160630.1 - 3 - 4 539 72
BC034361.1 - 4 - 5 1062 72

View Full Product Details