Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amz1
Entrez Gene ID
231842
Gene Name
archaelysin family metallopeptidase 1
Gene Aliases
5330426I05, 6530401C20Rik
Chromosome Location
Chr.5: 140724127 - 140753312 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173405.2 NP_775581.1 5 - 6 963 60
GenBank mRNA AJ879912.1 - 3 - 4 604 60
AK078332.1 - 5 - 6 963 60
AK155136.1 - - 850 60
AK170176.1 - 4 - 5 937 60
BC129841.1 - 3 - 4 746 60
BC129842.1 - 3 - 4 698 60

View Full Product Details