Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fnip2
Entrez Gene ID
329679
Gene Name
folliculin interacting protein 2
Gene Aliases
D630023B12Rik, mKIAA1450
Chromosome Location
Chr.3: 79455971 - 79567879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162999.2 NP_001156471.1 11 - 12 1360 92
GenBank mRNA AK122512.1 - 3 - 4 437 92
BC116316.1 - 4 - 5 624 92
BC116317.1 - 4 - 5 624 92

View Full Product Details