Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fnip2
Entrez Gene ID
329679
Gene Name
folliculin interacting protein 2
Gene Aliases
D630023B12Rik, mKIAA1450
Chromosome Location
Chr.3: 79455971 - 79567879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162999.2 NP_001156471.1 14 - 15 3008 61
GenBank mRNA AK122512.1 - 6 - 7 2085 61
AK141700.1 - 2 - 3 1129 61
BC116316.1 - 7 - 8 2272 61
BC116317.1 - 7 - 8 2272 61

View Full Product Details