Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc51
Entrez Gene ID
66658
Gene Name
coiled-coil domain containing 51
Gene Aliases
5730568A12Rik, AI551049
Chromosome Location
Chr.9: 109082496 - 109093363 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025689.4 NP_079965.2 3 - 4 541 69
GenBank mRNA AK014056.1 - 2 - 3 481 69
AK017860.1 - 3 - 4 538 69
AK137013.1 - 3 - 4 545 69
AK141240.1 - 3 - 4 544 69
BC098221.1 - 2 - 3 487 69
BC116786.1 - 2 - 3 399 69
BC116788.1 - 2 - 3 399 69

View Full Product Details