The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136064.3 | NP_001129536.1 | 4 - 5 | 1002 | 78 |
NM_001290823.1 | NP_001277752.1 | 3 - 4 | 461 | 78 | |
NM_008144.5 | NP_032170.3 | 3 - 4 | 852 | 78 | |
GenBank mRNA | AB030196.1 | - | 3 - 4 | 471 | 78 |
AF069954.2 | - | 3 - 4 | 677 | 78 | |
AK004634.1 | - | 1 - 2 | 2551 | 78 | |
AK151606.1 | - | 3 - 4 | 430 | 78 | |
AK155753.1 | - | 2 - 3 | 969 | 78 | |
AK158901.1 | - | 3 - 4 | 853 | 78 | |
AK170183.1 | - | 3 - 4 | 428 | 78 | |
BC043023.1 | - | 3 - 4 | 616 | 78 | |
BC060048.1 | - | 3 - 4 | 807 | 78 | |
BC061689.1 | - | 4 - 5 | 740 | 78 |