The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081071.2 | NP_001074540.1 | 5 - 6 | 826 | 62 |
NM_001177967.1 | NP_001171438.1 | 5 - 6 | 805 | 62 | |
NM_001177968.1 | NP_001171439.1 | 5 - 6 | 794 | 62 | |
GenBank mRNA | AK144580.1 | - | 5 - 6 | 769 | 62 |
BC147499.1 | - | 5 - 6 | 744 | 62 | |
BC150884.1 | - | 5 - 6 | 733 | 62 | |
BC158076.1 | - | 5 - 6 | 765 | 62 | |
BC158079.1 | - | 5 - 6 | 765 | 62 |