Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxw9
Entrez Gene ID
68628
Gene Name
F-box and WD-40 domain protein 9
Gene Aliases
1110017H11Rik, Fbw9
Chromosome Location
Chr.8: 85060025 - 85067121 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026791.2 NP_081067.2 8 - 9 1251 124
GenBank mRNA AK003747.1 - 3 - 4 315 124
AK082085.1 - 7 - 8 1740 124
AK156709.1 - 7 - 8 1401 124
BC043658.1 - 8 - 9 1212 124
BC053694.1 - 6 - 7 667 124
BC060082.1 - 5 - 6 482 124
BC144773.1 - 8 - 9 1617 124
BC145661.1 - 8 - 9 1617 124

View Full Product Details