The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166482.1 | NP_001159954.1 | 4 - 5 | 428 | 68 |
NM_172761.3 | NP_766349.1 | 4 - 5 | 522 | 68 | |
GenBank mRNA | AK083317.1 | - | 4 - 5 | 462 | 68 |
AK141278.1 | - | 4 - 5 | 357 | 68 | |
BC023782.1 | - | 4 - 5 | 499 | 68 | |
BC026652.1 | - | 3 - 4 | 398 | 68 | |
BC052066.1 | - | 4 - 5 | 428 | 68 | |
BQ444001.1 | - | 4 - 5 | 428 | 68 |