Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Taf4b
Entrez Gene ID
72504
Gene Name
TAF4B RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
105kDa, 2610524B04Rik, 4932409F03Rik, AW987595, TAFII-105, TAFII105, Taf2c2
Chromosome Location
Chr.18: 14783201 - 14960424 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100449.1 NP_001093919.1 5 - 6 1229 125

View Full Product Details