The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205236.1 | NP_001192165.1 | 15 - 16 | 2764 | 57 |
NM_133715.5 | NP_598476.2 | 13 - 14 | 1733 | 57 | |
GenBank mRNA | AK046763.1 | - | 13 - 14 | 1734 | 57 |
AK143916.1 | - | 11 - 12 | 1205 | 57 | |
AK154926.1 | - | 14 - 15 | 3055 | 57 | |
AK201966.1 | - | 1 - 2 | 58 | 57 |