Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Afap1l2
Entrez Gene ID
226250
Gene Name
actin filament associated protein 1-like 2
Gene Aliases
AU041783, C86904, mKIAA1914
Chromosome Location
Chr.19: 56912354 - 57008590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177796.1 NP_001171267.1 15 - 16 2254 84
NM_001177797.1 NP_001171268.1 14 - 15 2200 84
NM_146102.2 NP_666214.1 16 - 17 2306 84
GenBank mRNA AK048649.1 - 16 - 17 1960 84
AK087449.1 - 15 - 16 1890 84
AK220430.1 - 14 - 15 1823 84
BC031515.1 - 15 - 16 1856 84

View Full Product Details