The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112744.1 | NP_001106215.1 | 3 - 4 | 736 | 58 |
GenBank mRNA | AK042195.1 | - | 3 - 4 | 732 | 58 |
AK153708.1 | - | 3 - 4 | 897 | 58 | |
BC095966.1 | - | 1 - 2 | 33 | 58 | |
BC137847.1 | - | 3 - 4 | 777 | 58 | |
BC144970.1 | - | 3 - 4 | 777 | 58 |