Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl10
Entrez Gene ID
72096
Gene Name
methyltransferase like 10
Gene Aliases
2010208K18Rik
Chromosome Location
Chr.7: 132827232 - 132852880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028095.1 NP_082371.1 2 - 3 216 112
GenBank mRNA AK008476.1 - 2 - 3 201 112
BC038323.1 - 2 - 3 171 112
BC051506.1 - 2 - 3 191 112
BC096622.1 - 2 - 3 200 112
BC116375.2 - 2 - 3 214 112
BQ830246.1 - 2 - 3 241 112

View Full Product Details