The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025346.1 | NP_079622.1 | 2 - 3 | 493 | 91 |
GenBank mRNA | AK075599.1 | - | 2 - 3 | 493 | 91 |
BC016075.1 | - | 2 - 3 | 347 | 91 |