The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159525.1 | NP_001152997.1 | 4 - 5 | 516 | 70 |
NM_023041.3 | NP_075528.3 | 6 - 7 | 792 | 70 | |
GenBank mRNA | AK012785.1 | - | 6 - 7 | 775 | 70 |
AK028449.1 | - | 4 - 5 | 510 | 70 | |
AK029368.1 | - | 6 - 7 | 793 | 70 | |
AK075576.1 | - | 6 - 7 | 779 | 70 | |
AK170175.1 | - | 6 - 7 | 781 | 70 | |
AK182815.1 | - | 3 - 4 | 377 | 70 | |
AK210467.1 | - | 1 - 2 | 152 | 70 | |
BC012517.1 | - | 6 - 7 | 727 | 70 | |
BC019767.1 | - | 6 - 7 | 775 | 70 | |
CT010305.1 | - | 6 - 7 | 768 | 70 | |
Y09046.1 | - | 6 - 7 | 773 | 70 |