Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmbox1
Entrez Gene ID
219150
Gene Name
homeobox containing 1
Gene Aliases
AI451877, AI604847, F830020C16Rik
Chromosome Location
Chr.14: 64811600 - 64949906 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC051457.1 - 8 - 9 1364 82

View Full Product Details