Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hnrnpu
- Entrez Gene ID
-
51810
- Gene Name
- heterogeneous nuclear ribonucleoprotein U
- Gene Aliases
-
AA408410, AI256620, AL024194, AL024437, AW557595, C86794, Hnrpu, SAFA, Sp120, hnRNP U
- Chromosome Location
-
Chr.1: 178321108 - 178337810 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK029944.1 |
- |
14 - 15 |
2581 |
73 |
View Full Product Details