The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038701.2 | NP_001033790.1 | 3 - 4 | 285 | 87 |
NM_008071.3 | NP_032097.1 | 3 - 4 | 337 | 87 | |
GenBank mRNA | AK039070.1 | - | 3 - 4 | 307 | 87 |
AK083051.1 | - | 3 - 4 | 349 | 87 | |
AK083107.1 | - | 3 - 4 | 286 | 87 | |
AK147247.1 | - | 3 - 4 | 291 | 87 | |
U14420.1 | - | 3 - 4 | 318 | 87 |