Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hist1h3e
- Entrez Gene ID
-
319151
- Gene Name
- histone cluster 1, H3e
- Gene Aliases
-
H3-f, H3.1
- Chromosome Location
-
Chr.13: 23561896 - 23562365 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178205.2 |
NP_835512.1 |
1 - 1 |
350 |
97 |
GenBank mRNA |
BC127042.1 |
- |
1 - 1 |
343 |
97 |
- Gene Symbol
- Hist1h3f
- Entrez Gene ID
-
260423
- Gene Name
- histone cluster 1, H3f
- Gene Aliases
-
H3.2-221, Hist1
- Chromosome Location
-
Chr.13: 23544052 - 23544954 on Build GRCm38
- Gene Symbol
- Hist1h3h
- Entrez Gene ID
-
319152
- Gene Name
- histone cluster 1, H3h
- Gene Aliases
-
H3-291
- Chromosome Location
-
Chr.13: 21717628 - 21718115 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC107285.2 |
- |
1 - 1 |
348 |
97 |
View Full Product Details