Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pabpc1l
- Entrez Gene ID
-
381404
- Gene Name
- poly(A) binding protein, cytoplasmic 1-like
- Gene Aliases
-
1810053B01Rik, Epab
- Chromosome Location
-
Chr.2: 164024770 - 164050538 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK007853.1 |
- |
1 - 2 |
195 |
65 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG695649.1 |
- |
1 - 2 |
188 |
|
Chr. 2 - 164047401 - 164050499 [+] on Build GRCm38 |
65 |
GenBankmRNA |
BY707856.1 |
- |
1 - 2 |
193 |
|
Chr. 2 - 164047388 - 164050479 [+] on Build GRCm38 |
65 |
GenBankmRNA |
DV653886.1 |
- |
5 - 6 |
634 |
|
Chr. 2 - 164037120 - 164050143 [+] on Build GRCm38 |
65 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide |
Ywhab |
1300003C17Rik |
-28.8kb
|
Sense |
Chr. - 163995197 - 164018588 on Build GRCm38 |
Downstream |
translocase of outer mitochondrial membrane 34 |
Tomm34 |
2610100K07Rik, TOM34 |
3.0kb
|
Anti-sense |
Chr. - 164053538 - 164071177 on Build GRCm38 |
View Full Product Details