The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103171.1 | NP_001096641.1 | 31 - 32 | 6936 | 81 |
NM_010862.2 | NP_034992.2 | 32 - 33 | 6990 | 81 | |
NM_182698.2 | NP_874357.2 | 30 - 31 | 3174 | 81 | |
GenBank mRNA | AF053130.1 | - | 29 - 30 | 3142 | 81 |
AF144095.1 | - | 32 - 33 | 6990 | 81 | |
AY331132.1 | - | 31 - 32 | 6936 | 81 | |
AY331133.1 | - | 30 - 31 | 3174 | 81 |