Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il31
Entrez Gene ID
76399
Gene Name
interleukin 31
Gene Aliases
1700013B14Rik
Chromosome Location
Chr.5: 123480149 - 123482634 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029594.1 NP_083870.1 1 - 2 67 102
GenBank mRNA AY509149.1 - 1 - 2 67 102
BC117082.1 - 1 - 2 66 102
BC117084.1 - 1 - 2 66 102

View Full Product Details