Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Arhgap27
- Entrez Gene ID
-
544817
- Gene Name
- Rho GTPase activating protein 27
- Gene Aliases
-
2310069I04Rik, 5730442P18Rik, Gm21552, Sh3d20
- Chromosome Location
-
Chr.11: 103331484 - 103363692 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC048412.1 |
- |
|
200 |
|
Chr. - - [-] on Build |
76 |
View Full Product Details