Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLCE
Entrez Gene ID
101106592
Gene Name
glucuronic acid epimerase
Gene Aliases
-
Chromosome Location
Chr.7: 17203132 - 17320018 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010269.4 XP_004010318.1 3 - 4 997 79

View Full Product Details