Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBP3
Entrez Gene ID
101115179
Gene Name
retinol binding protein 3
Gene Aliases
IRBP
Chromosome Location
Chr.25: 44064972 - 44074616 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021549.4 XP_004021598.2 3 - 4 3546 91

View Full Product Details