Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BSCL2
Entrez Gene ID
101113634
Gene Name
BSCL2 lipid droplet biogenesis associated, seipin
Gene Aliases
-
Chromosome Location
Chr.21: 42877670 - 42893152 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960099.1 XP_027815900.1 9 - 10 1283 89

View Full Product Details