Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSC
Entrez Gene ID
702308
Gene Name
goosecoid homeobox
Gene Aliases
-
Chromosome Location
Chr.7: 156947510 - 156949546 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100695.4 XP_001100695.1 - 859 64

View Full Product Details