The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015133200.2 | XP_014988686.1 | - | 742 | 105 |
XM_028845522.1 | XP_028701355.1 | - | 365 | 105 | |
XM_028845523.1 | XP_028701356.1 | - | 448 | 105 | |
GenBank mRNA | JU327438.1 | - | - | 499 | 105 |
JV465160.1 | - | - | 492 | 105 |